Skip to Content
Merck

B3936

BAY R3401

≥98% (HPLC), solid

Synonym(s):

4-(2-Chlorophenyl)-1-ethyl-2-methyl-5-oxo-1,4,5,7-tetrahydrofuro[3,4-b]pyridine-3-carboxylic acid isopropyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H22ClNO4
CAS Number:
Molecular Weight:
375.85
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

solid

color

white

solubility

DMSO: 15 mg/mL

originator

Bayer

storage temp.

2-8°C

SMILES string

CCN1C(C)=C(C(c2ccccc2Cl)C3=C1COC3=O)C(=O)OC(C)C

InChI

1S/C20H22ClNO4/c1-5-22-12(4)16(20(24)26-11(2)3)17(13-8-6-7-9-14(13)21)18-15(22)10-25-19(18)23/h6-9,11,17H,5,10H2,1-4H3

InChI key

NPUSXSOBPNHOPH-UHFFFAOYSA-N

Biochem/physiol Actions

BAY R3401 is a glycogen phosphorylase inhibitor. The racemic prodrug, BAY R3401, suppresses hepatic glycogenolysis. BAY W1807, the active metabolite of BAY R3401, inhibits muscle glycogen phosphorylase a and b. It has been investigated that the metabolites of BAY R3401 suppress hepatic glycogenolysis by allosteric inhibition and dephosphorylation of phosphorylase a.
Glycogen phosphorylase inhibitor.

Features and Benefits

This compound was developed by Bayer. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Packaging

Light sensitive; store in amber vials.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library