2051725
Direct
pkg of 25 ea
About This Item
Productos recomendados
packaging
pkg of 25 ea
L × O.D. × I.D.
78.5 mm × 6.3 mm × 1.5 mm
General description
Inlet liner is an important part within the injection port. The prime functions of inlet liner are to limit sample degradation and improve vaporisation along with correctly positioning the syringe needle. A wrong choice of liner can have prominent effect on the chromatogram. The basic problems related with incorrect choice of liner include the higher potential of peak tailing and mass discrimination. The inlet liner volume is recommended to be larger than the amount of sample injected by syringe so as to avoid any reaction with the sample. Injector port liner is suitable to make connection between sample introduction and GC column. Direct type has narrow bore which sustains a high linear velocity through the injection port to diminish band broadening and transferring all the samples to column.
Deactivated glass liner for capillary injection port
Application
Inlet liner, direct type may be used for solid phase microextraction (SPME) sample injection.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Lo sentimos, en este momento no disponemos de COAs para este producto en línea.
¿Ya tiene este producto?
Novel aggregation pheromone,(1S, 4R)-p-menth-2-en-1-ol, of the ambrosia beetle, Platypus quercivorus (Coleoptera: Platypodidae)
Bulletin of the Forestry and Forest Products Research Institute, 6, 49-57 (2007)
Forensic Applications of Gas Chromatography, 9-9 (2013)