Z694126
Tygon® 2475 high purity tubing
About This Item
Código UNSPSC:
41105108
NACRES:
NB.35
Productos recomendados
envase
pack of 50 feet
fabricante / nombre comercial
Saint Gobain ACG00017
D.I. × D.E.
1/4 in. × 3/8 in. × 1/16 in.
Descripción general
TYGON® 2475 high-purity tubing is a plasticizer-free tubing that utilizes the latest in polymer technology to provide an entirely clear and flexible tubing choice for sensitive fluid transfer applications. TYGON 2475 is hydrophobic and will resist the absorption/adsorption of aqueous fluids. This reduction in sorption minimizes the risk of fluid alteration in single- or repeat-use applications. The smooth inner surface of TYGON 2475 inhibits particulate entrapment on the tubing wall and reduces the potential for bacterial growth and contamination.
Características y beneficios
- Exceptionally low absorption/adsorption compared to silicone
- Smooth inner surface inhibits particulate entrapment
- Environmentally safe to dispose and reduces disposal costs
- Highly resistant to aggressive cleaners and sanitizers
- Plasticizer-free
- Meets all USP Class VI and FDA criteria
Typical Applications:
- Sterile filling and dispensing systems
- Diagnostic equipment
- Laboratory analytical instrumentation
- Infusion sets for parenterals and drugs
- Cell and tissue culture transport
Información legal
Tygon is a registered trademark of Saint-Gobain Performance Plastics Corp.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)