BMSW1105131000
Accuris™ M1 Grade Calibration Weight Set
pack of 13 pieces, size 1-1000 g, stainless steel
About This Item
Código UNSPSC:
41100000
NACRES:
NB.71
Productos recomendados
Materiales
stainless steel
envase
pack of 13 pieces
fabricante / nombre comercial
Benchmark Scientific W1105-13100
tamaño
1-1000 g
Categorías relacionadas
Descripción general
Basic, compact balances with readability in the range of 0.1g to 0.01g can be calibrated with M1 Class weights.
Each individual weight and weight set from Accuris™ is supplied with an individual Certificate of Verification confirming that the weight(s) meet OIML regulations and tolerances for the specific weight class.
Each individual weight and weight set from Accuris™ is supplied with an individual Certificate of Verification confirming that the weight(s) meet OIML regulations and tolerances for the specific weight class.
The economy M1 Calibration Weight Set includes 13 pieces: 1 x 1g, 2x2g, 1x5g, 1x10g, 2x20g, 1x50g, 1x100g, 2x200g, 1x500g and 1x1Kg. A protective carrying case is included.
Información legal
Accuris is a trademark of Benchmark Scientific, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)