Key Documents
Z177350
Scienceware® micro funnel
top I.D. 28 mm, stem diam. 6 mm
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
packaging
pack of 12 ea
manufacturer/tradename
Bel-Art F14685-0028
stem diam.
6 mm
top I.D.
28 mm
Categorías relacionadas
General description
PP. Useful for micro work, filling burettes, manometers, etc.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP