Skip to Content
Merck
All Photos(1)

Key Documents

5.06045

Sigma-Aldrich

GPR55 Receptor Antagonist, CID16020046

Synonym(s):

GPR55 Receptor Antagonist, CID16020046, 4-[4-(3-Hydroxyphenyl)-3-(4-methylphenyl)-6-oxo-1H,4H-pyrrolo[3,4-c]pyrazol-5-yl]benzoic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H19N3O4
CAS Number:
Molecular Weight:
425.44
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥99% (HPLC)

Quality Level

form

powder

potency

1 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

CC1=CC=C(C=C1)C2=NNC3=C2C(N(C3=O)C4=CC=C(C=C4)C(=O)O)C5=CC(=CC=C5)O

InChI

1S/C25H19N3O4/c1-14-5-7-15(8-6-14)21-20-22(27-26-21)24(30)28(18-11-9-16(10-12-18)25(31)32)23(20)17-3-2-4-19(29)13-17/h2-13,23,29H,1H3,(H,26,27)(H,31,32)

General description

A selective antagonist of G protein-coupled receptor 55 (GPR55) that blocks lipid L-a-lysophosphatidylinositol (LPI, Cat. No. 440153)-induced GPR55 activation (IC50 = 1 µM) and downstream effects in HEK293 cells. Also shown to block the agonistic effect of LPI on GPR55- expressing yeast cells (~10 µM). Inhibits LPI-induced Ca2+ mobilization and ERK1/2 phosphorylation in HEK-GPR55 and HEK-CB1 cells. However, it does not affect basal Ca2+ mobilization and ERK1/2 activity. Blocks the GPR55-mediated NFAT and NF-kB activation and their nuclear translocation in HEK-GPR55 cells. Diminishes LPI-induced wound healing in GPR55-expressing primary human lung microvascular endothelial cells and reverses LPI-inhibited platelet aggregation (~10 µM). Also reported to inhibit the activity of phosphodiesterases PDE3A and PDE4B (pIC50 = 5 and 4.8, respectively).

Biochem/physiol Actions

Primary Target
GPR55

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kargl, J., et al, 2013. J. Pharmacol. Exp. Ther.346, 54.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service