Skip to Content
Merck
All Photos(1)

Key Documents

SAB1410929

Sigma-Aldrich

Anti-SNX9 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

MST155, MSTP155, SDP1, SH3PX1, SH3PXD3A, WISP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 66.6 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SNX9(51429)

General description

This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like some family members, but does contain a SH3 domain near its N-terminus. This protein interacts with the cytoplasmic domains of the precursor but not the processed forms of a disintegrin and metalloprotease domain 9 and 15. This protein binds the beta-appendage domain of adaptor protein 2 and may function to assist adaptor protein 2 in its role at the plasma membrane. This protein interacts with activated Cdc42-associated kinase-2 to regulate the degradation of epidermal growth factor receptor protein. (provided by RefSeq)

Immunogen

SNX9 (NP_057308.1, 1 a.a. ~ 595 a.a) full-length human protein.

Sequence
MATKARVMYDFAAEPGNNELTVNEGEIITITNPDVGGGWLEGRNIKGERGLVPTDYVEILPSDGKDQFSCGNSVADQAFLDSLSASTAQASSSAASNNHQVGSGNDPWSAWSASKSGNWESSEGWGAQPEGAGAQRNTNTPNNWDTAFGHPQAYQGPATGDDDDWDEDWDGPKSSSYFKDSESADAGGAQRGNSRASSSSMKIPLNKFPGFAKPGTEQYLLAKQLAKPKEKIPIIVGDYGPMWVYPTSTFDCVVADPRKGSKMYGLKSYIEYQLTPTNTNRSVNHRYKHFDWLYERLLVKFGSAIPIPSLPDKQVTGRFEEEFIKMRMERLQAWMTRMCRHPVISESEVFQQFLNFRDEKEWKTGKRKAERDELAGVMIFSTMEPEAPDLDLVEIEQKCEAVGKFTKAMDDGVKELLTVGQEHWKRCTGPLPKEYQKIGKALQSLATVFSSSGYQGETDLNDAITEAGKTYEEIASLVAEQPKKDLHFLMECNHEYKGFLGCFPDIIGTHKGAIEKVKESDKLVATSKITLQDKQNMVKRVSIMSYALQAEMNHFHSNRIYDYNSVIRLYLEQQVQFYETIAEKLRQALSRFPVM

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service