CLS2982100
Pyrex® graduated cylinders with Pyrex® stopper
volume 100 mL
Synonym(s):
Corning® 2982
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121805
NACRES:
NB.43
Recommended Products
feature
subdivision 1.0 mL
manufacturer/tradename
Corning 2982-100
tol. measuring range
0.6
volume
100 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Single metric scale
- Calibrated "to contain"
- White enamel graduations
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service