Skip to Content
Merck
All Photos(1)

Key Documents

445509

Sigma-Aldrich

2-Allyloxybenzaldehyde

technical grade

Synonym(s):

2-(2-Propenyloxy)benzaldehyde, 2-(Allyloxy)benzaldehyde, o -(Allyloxy)benzaldehyde, o -Allylsalicylaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCH2OC6H4CHO
CAS Number:
Molecular Weight:
162.19
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

form

liquid

refractive index

n20/D 1.55 (lit.)

bp

130 °C/10 mmHg (lit.)

density

1.079 g/mL at 25 °C (lit.)

SMILES string

C=CCOc1ccccc1C=O

InChI

1S/C10H10O2/c1-2-7-12-10-6-4-3-5-9(10)8-11/h2-6,8H,1,7H2

InChI key

BXCJDECTRRMSCV-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service