Skip to Content
Merck
All Photos(1)

Key Documents

567051-M

Millipore

Sinefungin

InSolution, ≥95%, antibiotic

Synonym(s):

InSolution Sinefungin, SF, Adenosyl-ornithine, A9145

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23N7O5
CAS Number:
Molecular Weight:
381.39
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

storage temp.

−20°C

SMILES string

[n]2(c3ncnc(c3nc2)N)[C@@H]1O[C@@H]([C@H]([C@H]1O)O)C[C@@H](N)CC[C@H](N)C(=O)O

InChI

1S/C15H23N7O5/c16-6(1-2-7(17)15(25)26)3-8-10(23)11(24)14(27-8)22-5-21-9-12(18)19-4-20-13(9)22/h4-8,10-11,14,23-24H,1-3,16-17H2,(H,25,26)(H2,18,19,20)/t6-,7-,8+,10+,11+,14+/m0/s1

InChI key

LMXOHSDXUQEUSF-YECHIGJVSA-N

General description

An anti-leishmanial nucleoside antibiotic that acts as an S-adenosyl-L-methionine (SAM, AdoMet) methyltransferase-specific inhibitor.

Biochem/physiol Actions

Primary Target
S-adenosyl-L-methionine (SAM, AdoMet) methyltransferase

Warning

Toxicity: Standard Handling (A)

Physical form

A 10 mM (2 mg/524 µl) solution in H₂O.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Cheng, D., et al. 2004. J. Biol. Chem.279, 23892.
Lin, Q., et al. 2001. J. Am. Chem. Soc.123, 11608.
Zingg, J.M., et al. 1996. Nucleic Acids Res.24, 3267.
McCammon, M.T., and Parks, L.W. 1981. J. Bacteriol.145, 106.
Pugh, C.S.G., et al. 1978. J. Biol. Chem.253, 4075.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service