GF11161066
T1 - thermocouple alloy, Ni90Cr10
microfoil, 25mm disks, 1μm thinness, permanent mylar 3.5μm support
Synonym(s):
NI094850
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11101711
NACRES:
NA.23
disc size
25 mm
description
3.5μm permanent mylar support
form
disc (microfoil)
manufacturer/tradename
Goodfellow 111-610-66
thickness
1 μm , 3.5μm permanent mylar support
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service