CLS556402L
Pyrex® class A low-actinic volumetric flask, Pyrex® ST stopper
capacity 2,000 mL
Synonym(s):
flasks glass, volumetric flasks, volumetric glass
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Recommended Products
material
borosilicate glass
long
Joint
joint stopper27
sterility
non-sterile
feature
class A
closure type ST stopper
packaging
case of 1
manufacturer/tradename
Corning 55640-2L
Corning 55640
tol. measuring range
0.5 mL
capacity
2,000 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Low actinic stained glass provides protection for materials sensitive to light. The protective color is an integral part of flask, which retains mechanical strength, chemical stability, and thermal resistance of Pyrex brand labware.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service