Skip to Content
Merck
All Photos(1)

Key Documents

674850

Sigma-Aldrich

4-Phenyl-2-(trifluoromethyl)butanoic acid

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11F3O2
Molecular Weight:
232.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

95%

form

solid

mp

55-61 °C

functional group

carboxylic acid
fluoro
phenyl

SMILES string

OC(=O)C(CCc1ccccc1)C(F)(F)F

InChI

1S/C11H11F3O2/c12-11(13,14)9(10(15)16)7-6-8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H,15,16)

InChI key

ILPBLRFRNISTEM-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service