605211
Thrombin Substrate III, Fluorogenic
Sensitive fluorogenic substrate for the quantitative determination of thrombin.
Synonym(s):
Thrombin Substrate III, Fluorogenic, Benzoyl-FVR-AMC
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C37H43N7O6 · xHCl
Molecular Weight:
681.78 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
lyophilized
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated
solubility
DMSO: 5 mg/mL
fluorescence
λex 360-380 nm
λem 440-460 nm
shipped in
ambient
storage temp.
−20°C
General description
Sensitive fluorogenic substrate for the quantitative determination of thrombin. Excitation max.: 360-380 nm; emission max.: 440-460 nm.
Warning
Toxicity: Standard Handling (A)
Sequence
Benzoyl-Phe-Val-Arg-AMC,HCl
Physical form
Supplied as a hydrochloride salt.
Reconstitution
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Other Notes
Tran, T. and Stewart, A. 2003. Br. J Pharmacol.138, 865.
Lottenberg, R., et al. 1981. Methods Enzymol. 80, 341.
Svendsen, L., et al. 1972. Folia Haematol.98, 446.
Lottenberg, R., et al. 1981. Methods Enzymol. 80, 341.
Svendsen, L., et al. 1972. Folia Haematol.98, 446.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service