BCR599
Ewes′/Goats’ Curd (for adulteration with cows’ milk)
0 and 1 % cows’ milk, BCR®, certified reference material
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
85151701
NACRES:
NA.24
Recommended Products
Product Name
Ewes′/Goats′ Curd (for adulteration with cows′ milk) 0 and 1 % cows′ milk, BCR®, certified reference material
grade
certified reference material
Agency
BCR®
manufacturer/tradename
JRC
application(s)
agriculture
food and beverages
format
matrix material
storage temp.
−20°C
Analysis Note
For more information please see:
BCR599
BCR599
Other Notes
Sales restrictions may apply
Legal Information
BCR is a registered trademark of European Commission
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service