Skip to Content
Merck
All Photos(1)

Key Documents

SLW3050/14M

Pyrex® Sintered glass filter disc

nominal diameter 50 mm, porosity grade 4

Synonym(s):

Pyrex, SciLabware 3050/14M

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
NACRES:
NB.24
pore size:
grade 4 porosity
material:
Pyrex membrane
nominal diameter:
50 mm
sterility:
non-sterile

material

Pyrex membrane

sterility

non-sterile

manufacturer/tradename

SciLabware 3050/14M

nominal diameter

50 mm

pore size

grade 4 porosity

Looking for similar products? Visit Product Comparison Guide

General description

  • Complies with ISO 4793
  • For use by Glassblowers constructing special apparatus
  • Manufactured from Pyrex borosilicate glass for resistance to heat and chemical corrosion
  • Close control of pore size
  • Accurate and consistent disc dimensions
  • High mechanical strength to withstand full vacuum
  • Convenient packaging

Legal Information

PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service