Merck
  • The transcription factor GATA1 regulates NBEAL2 expression through a long-distance enhancer.
The transcription factor GATA1 regulates NBEAL2 expression through a long-distance enhancer.
Haematologica (2017-01-14)
Anouck Wijgaerts, Christine Wittevrongel, Chantal Thys, Timothy Devos, Kathelijne Peerlinck, Marloes R Tijssen, Chris Van Geet, Kathleen Freson
ABSTRACT
Gray platelet syndrome is named after the gray appearance of platelets due to the absence of α-granules. It is caused by recessive mutations in
Sigma-Aldrich
MISSION® esiRNA, targeting human GATA1
Pricing and availability is not currently available.