Merck
  • Regulation of NKG2D-ligand cell surface expression by intracellular calcium after HDAC-inhibitor treatment.
Regulation of NKG2D-ligand cell surface expression by intracellular calcium after HDAC-inhibitor treatment.
Molecular immunology (2012-09-12)
Helle Jensen, Michael Hagemann-Jensen, Felicia Lauridsen, Søren Skov
ABSTRACT
In this study we demonstrate that histone deacetylase (HDAC)-inhibitor mediated cell surface expression of the structural different NKG2D-ligands MICA/B and ULBP2 is calcium-dependent. Treatment with the calcium chelator EGTA inhibited constitutive as well as HDAC-inhibitor induced MICA/B and ULBP2 cell surface expression on melanoma cells and Jurkat T-cells. A NKG2D-dependent cytolytic assay and staining with a recombinant NKG2D-Fc fusion protein showed that calcium chelation impaired the functional ability of NKG2D-ligands induced by HDAC-inhibitor treatment. The HDAC-inhibitor induced cell surface expression of ULBP2, but not MICA/B, was sensitive to treatment calmidazolium and trifluoperazine, two agents known to block calcium signaling. siRNA-mediated knock-down of the calcium-regulated proteins calmodulin or calpain did however not affect NKG2D-ligand cell surface expression on Jurkat T-cells. We further show that secretion and cell surface binding of the calcium-regulating protein galectin-1 is enhanced upon HDAC-inhibitor treatment of melanoma cells. However, binding of galectin-1 to cell surface glycoproteins was not critical for constitutive or HDAC-inhibitor induced MICA/B and ULBP2 cell surface expression. We provide evidence that MICA/B and ULBP2 cell surface expression is controlled differently by calcium, which adds to the increasing perception that cell surface expression of MICA/B and ULBP2 is controlled by distinct signal transduction pathways.
Sigma-Aldrich
Calmidazolium chloride, solid