- LuxR dependent quorum sensing inhibition by N,N'-disubstituted imidazolium salts.
LuxR dependent quorum sensing inhibition by N,N'-disubstituted imidazolium salts.
Bioorganic & medicinal chemistry (2011-07-26)
Mohamad Sabbah, Laurent Soulère, Sylvie Reverchon, Yves Queneau, Alain Doutheau
PMID21782453
ABSTRACT
Thirty N,N'-disubstituted imidazolium salts have been synthesized and evaluated as LuxR antagonists. Substitution on one of the imidazolium nitrogen atoms includes benzhydryl, fluorenyl or cyclopentyl substituent, and alkyl chains of various lengths on the second one. Most of these compounds displayed antagonist activity, with IC(50) reaching the micromolar range for the most active ones. The disubstituted imidazolium scaffold is thus shown to be a new pertinent pharmacophore in the field of AHL dependent QS inhibition.