
Lauroyl-L-carnitine

Synonym(s):
(2R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxododecyl)oxy]-1-propanaminium inner salt, L-Carnitine dodecanoyl ester, C12-Carnitine, Dodecanoyl-L-carnitine
Empirical Formula (Hill Notation):
C19H37NO4
CAS Number:
Molecular Weight:
343.50
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Application
Optical Isomer
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing