713-0922
HPLC Analysis of Ferredoxin NADP Reductase on TSKgel® Phenyl-5PW
application for HPLC
About This Item
technique(s)
HPLC: suitable
test parameters
column: TSKgel Phenyl-5PW column, 7.5 cm × 7.5 mm I.D., 10 μm particles (807573)
mobile phase: 60 min decreasing linear gradient from 1.5M to 0M ammonium sulfate in 0.1M sodium phosphate buffer, pH7
flow rate: 0.5 mL/min
column temp.: 25 °C
detector: UV, 280 nm
injection: 3mg (crude) or 0.2mg (standard)
suitability
application for HPLC
application(s)
life science and biopharma
Legal Information
Analyte
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service