GBL645502
Grace Bio-Labs CoverWell™ incubation chambers
chamber volume 500 μL, W × L × thickness 22 mm × 40 mm × 0.5 mm
Synonym(s):
leak proof chamber, leak proof enclosure, microscope stick on chamber, reusable press to seal chamber
About This Item
Recommended Products
material
red
silicone
sterility
non-sterile
packaging
pack of 50 ea
manufacturer/tradename
Grace Bio-Labs 645502
W × L × thickness
22 mm × 40 mm × 0.5 mm
chamber volume
500 μL
wells
1
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Eliminate precipitate deposits on specimens by incubating slides and specimens upside down during enzymatic color precipitation reaction.
- Provides an exceptionally secure seal during submerged water bath and/or high temperature incubations.
- RNase and DNase free.
- Hydrophobic.
- Adheres to wet or dry surfaces.
- Reusable.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service