Skip to Content
Merck
All Photos(1)

Documents

BAF420740220

SP Bel-Art® Secador® 4.0 Horizontal Auto-Desiccator Cabinet

internal volume 1.9 ft3, AC/DC input 220 V / 240 V, universal plug set, with Clear Door, clear polyester (Co-polyester)

Synonym(s):

SECADOR 4.0 HORIZONTAL AUTO-DESICCATOR CABINETS, SP BEL-ART SECADOR CLEAR 4.0 HORIZONTAL AUTO-DESICCATOR CABINET; 230V, 1.9 CU. FT.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104510
NACRES:
NB.77

material

clear polyester (Co-polyester)
polycarbonate

feature

with Clear Door

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art F42074-0220

internal volume

1.9 ft3

Looking for similar products? Visit Product Comparison Guide

General description

Protecting valuable humidity-sensitive items, such as reagents or electronics, is easy and fully automatic with Secador® Auto-Desiccator Cabinets. The patented, electronic desiccant regeneration module recharges the desiccant every 20 minutes. Compared to other auto-desiccators that regenerate once every 4 to 6 hours, the Secador® Auto-Desiccator quickly attains a low %rH level on start-up and consistently maintains low %rH, especially with frequent entry to the desiccator interior.
  • Polycarbonate construction blocks 99% of UV light and resists staining, crazing and chemical attack
  • Amber color models reduce visible light penetration by over 50%; perfect for storing light sensitive materials like reagents, analytical standards and investigational compounds
  • Color-indicating silica gel inside the desiccant regeneration module allows quick visual confirmation of desiccant status
  • Large, pull down, oven style door with easy-to-view digital hygrometer maximizes access to interior space; includes patented seal design, tab latches and security loops for a padlock or tamper-evident seals; opens right to left
  • Two removable, perforated shelves facilitate circulation of dry air
  • Choose from a selection of sizes, styles and colors in 100, 120 or 230V (CE marked) models for use worldwide
  • 230V units include power cord/transformer that includes Q-NA, Q-SAA, Q-UK and Q-EU input connectors to accommodate North American, Australian, UK and European configurations
  • Replacement shelves available; 4.0 units can be converted from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves
  • Use BAF420740020 replacement desiccant for Secador® Auto Desiccator Module

Legal Information

Bel-Art is a registered trademark of SP Scienceware, a division of SP
Secador is a registered trademark of SP Scienceware, a division of SP

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service