Merck
  • Validation of Computational Studies for Electrical Brain Stimulation With Phantom Head Experiments.
Validation of Computational Studies for Electrical Brain Stimulation With Phantom Head Experiments.
Brain stimulation (2015-07-26)
Donghyeon Kim, Jinmo Jeong, Sangdo Jeong, Sohee Kim, Sung Chan Jun, Euiheon Chung
ABSTRACT
Although computational studies of electrical brain stimulation (EBS) have received attention as a cost-effective tool, few studies have validated the technique, particularly in invasive cortical stimulation. In order to validate such studies, we used EBS to compare electric potential distributions generated by both numerical simulations and empirical measurements in three phantom head models (one-/three-layered spherical heads and MRI-based head). We constructed spherical phantom heads that consisted of one or three layers, and an anatomical, MRI-based phantom that consisted of three layers and represented the brain or brain/skull/scalp in order to perform both numerical simulations using the finite element method (FEM) and experimental measurements. Two stimulation electrodes (cathode and anode) were implanted in the phantoms to inject regulated input voltage, and the electric potential distributions induced were measured at various points located either on the surface or deep within the phantoms. We observed that both the electric potential distributions from the numerical simulations and experiments behaved similarly and resulted in average relative differences of 5.4% (spherical phantom) and 10.3% (MRI-based phantom). This study demonstrated that numerical simulation is reasonably consistent with actual experimental measurements; thus, because of its cost-effectiveness, EBS computational studies may be an attractive approach for necessary intensive/extensive studies.
Sigma-Aldrich
Agarose, High EEO, for molecular biology
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, low EEO
Sigma-Aldrich
Agarose, Special High EEO
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature, molecular biology grade
Sigma-Aldrich
Agarose, For pulsed field electrophoresis running gel
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, Wide range/Standard 3:1
Sigma-Aldrich
Agarose, Type I-A, low EEO
Sigma-Aldrich
Agarose, Type I, low EEO
Sigma-Aldrich
Sepharose 6B, 6% Beaded Agarose, 45-165 μm (wet), fractionation range 10,000-1,000,000 Da (dextrans)
Sigma-Aldrich
Agarose, High EEO
Sigma-Aldrich
Agarose, Low EEO
Sigma-Aldrich
Agarose, for molecular biology
Sepharose 6B, Cytiva 17-0110-01, pack of 1 L
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature
Sigma-Aldrich
Agarose, High Gelling Temperature
Sigma-Aldrich
Agarose, Wide range, for molecular biology
Sigma-Aldrich
Agarose, Type IV, Special High EEO
Sigma-Aldrich
Agarose, Medium EEO, for molecular biology
Sigma-Aldrich
Agarose, Low EEO, for Immunoelectrophoresis
Sigma-Aldrich
Agarose, Type II-A, Medium EEO
Sepharose 4B, Cytiva 17-0120-01, pack of 1 L
Sigma-Aldrich
Sepharose 4B, 45-165 μm bead diameter