Merck
  • Pharmacokinetics and preventive effects of platinum nanoparticles as reactive oxygen species scavengers on hepatic ischemia/reperfusion injury in mice.
Pharmacokinetics and preventive effects of platinum nanoparticles as reactive oxygen species scavengers on hepatic ischemia/reperfusion injury in mice.
Metallomics : integrated biometal science (2014-03-25)
Hidemasa Katsumi, Kentaro Fukui, Kanako Sato, Shoko Maruyama, Shugo Yamashita, Erika Mizumoto, Kosuke Kusamori, Munetaka Oyama, Masataka Sano, Toshiyasu Sakane, Akira Yamamoto
ABSTRACT
Reactive oxygen species (ROS) are involved in the pathophysiology of ischemia/reperfusion injury. To protect mouse hepatocytes from ischemia/reperfusion injury, we prepared two different sizes of citric acid-protected platinum nanoparticles (Pt-NPs), which exhibited ROS-scavenging activities and selective delivery to a specific type of liver cell. Small Pt-NPs (30 nm) reduced the superoxide anion, hydrogen peroxide, and hydroxyl radical levels in solution to a greater extent than did large Pt-NPs (106 nm). Large and small Pt-NPs predominantly accumulated in hepatic nonparenchymal cells after intravenous injection into mice. In a mouse model of ischemia/reperfusion injury, in which hepatic injury was induced by occluding the portal vein for 15 min followed by 6 h reperfusion, the increase in plasma alanine aminotransferase (ALT) and aspartate aminotransferase (AST) activities was inhibited by a bolus intravenous injection of either large or small Pt-NPs. However, small Pt-NPs inhibited the increase in these markers of hepatic injury to a greater extent than did large Pt-NPs. These results indicate that Pt-NPs can be used to prevent hepatic ischemia/reperfusion injury. To our knowledge, this is the first report demonstrating the pharmacokinetics and efficacy of Pt-NPs to prevent hepatic ischemia/reperfusion injury.
Sigma-Aldrich
Quantification kit for oxidized and reduced glutathione, 200 tests
Pricing and availability is not currently available.