Merck
  • Effect of eculizumab on hemolysis and transfusion requirements in patients with paroxysmal nocturnal hemoglobinuria.
Effect of eculizumab on hemolysis and transfusion requirements in patients with paroxysmal nocturnal hemoglobinuria.
The New England journal of medicine (2004-02-06)
Peter Hillmen, Claire Hall, Judith C W Marsh, Modupe Elebute, Michael P Bombara, Beth E Petro, Matthew J Cullen, Stephen J Richards, Scott A Rollins, Christopher F Mojcik, Russell P Rother
ABSTRACT
Paroxysmal nocturnal hemoglobinuria (PNH) arises from a somatic mutation of the PIG-A gene in a hematopoietic stem cell and the subsequent production of blood cells with a deficiency of surface proteins that protect the cells against attack by the complement system. We tested the clinical efficacy of eculizumab, a humanized antibody that inhibits the activation of terminal complement components, in patients with PNH. Eleven transfusion-dependent patients with PNH received infusions of eculizumab (600 mg) every week for four weeks, followed one week later by a 900-mg dose and then by 900 mg every other week through week 12. Clinical and biochemical indicators of hemolysis were measured throughout the trial. Mean lactate dehydrogenase levels decreased from 3111 IU per liter before treatment to 594 IU per liter during treatment (P=0.002). The mean percentage of PNH type III erythrocytes increased from 36.7 percent of the total erythrocyte population to 59.2 percent (P=0.005). The mean and median transfusion rates decreased from 2.1 and 1.8 units per patient per month to 0.6 and 0.0 units per patient per month, respectively (P=0.003 for the comparison of the median rates). Episodes of hemoglobinuria were reduced by 96 percent (P<0.001), and measurements of the quality of life improved significantly. Eculizumab is safe and well tolerated in patients with PNH. This antibody against terminal complement protein C5 reduces intravascular hemolysis, hemoglobinuria, and the need for transfusion, with an associated improvement in the quality of life in patients with PNH.
Sigma-Aldrich
Complement C5 from human serum, ≥90% (SDS-PAGE), >100,000 C5H50 units/mg protein
Pricing and availability is not currently available.