Skip to Content
Merck
All Photos(1)

Documents

V800158

Sigma-Aldrich

Diethyl phthalate

LR, ≥99%

Synonym(s):

Anozol, Diethyl benzene-1,2-dicarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(CO2C2H5)2
CAS Number:
Molecular Weight:
222.24
Beilstein:
1912500
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:

grade

LR

vapor density

7.66 (vs air)

vapor pressure

1 mmHg ( 100 °C)

product line

Vetec

Assay

≥99%

form

viscous liquid

autoignition temp.

854 °F

expl. lim.

0.75 %, 187 °F

refractive index

n20/D 1.502 (lit.)

bp

298-299 °C (lit.)

mp

−3 °C (lit.)

density

1.12 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)c1ccccc1C(=O)OCC

InChI

1S/C12H14O4/c1-3-15-11(13)9-7-5-6-8-10(9)12(14)16-4-2/h5-8H,3-4H2,1-2H3

InChI key

FLKPEMZONWLCSK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

338.0 °F - closed cup

Flash Point(C)

170 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lee-Ju Cheng et al.
Aquatic toxicology (Amsterdam, Netherlands), 109, 166-175 (2011-10-28)
The toxicity and effects of diethyl phthalate (DEP), a potent allelochemical, on the growth of greater duckweed were studied. Biochemical analyses and physiological methods were combined to investigate oxidative stress, adverse effects and their mechanisms in greater duckweeds grown in
Maik Hadorn et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(50), 20320-20325 (2012-11-24)
Higher-order structures that originate from the specific and reversible DNA-directed self-assembly of microscopic building blocks hold great promise for future technologies. Here, we functionalized biotinylated soft colloid oil-in-water emulsion droplets with biotinylated single-stranded DNA oligonucleotides using streptavidin as an intermediary
Seungmin Na et al.
Ultrasonics sonochemistry, 19(5), 1094-1098 (2012-04-03)
This study investigated the degradation of diethyl phthalate (DEP) by sonolytic, photolytic and sonophotolytic processes. Two types of UV lamps, UVC (254 nm) and VUV (185 nm+254 nm), were combined with ultrasound (283 kHz). The pseudo-first order degradation rate constants
Raphael J Witorsch et al.
Critical reviews in toxicology, 40 Suppl 3, 1-30 (2010-10-12)
This article reviews laboratory and epidemiological research into the endocrine disruptive effects of components of personal care products, namely, phthalate esters, parabens, ultraviolet (UV) filters, polycyclic musks, and antimicrobials. High doses of phthalates in utero can produce “phthalate syndrome,” demasculinizing
Kashappa-Goud H Desai et al.
Journal of controlled release : official journal of the Controlled Release Society, 165(1), 62-74 (2012-10-30)
Herein, we describe the detailed development of a simple and effective method to microencapsulate vaccine antigens in poly(lactic-co-glycolic acid) (PLGA) by simple mixing of preformed active self-microencapsulating (SM) PLGA microspheres in a low concentration aqueous antigen solution at modest temperature

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service