Iupac Name
N-[2-(1H-indol-3-yl)ethyl]nicotinamide
Inchi Code
1S/C16H15N3O/c20-16(13-4-3-8-17-10-13)18-9-7-12-11-19-15-6-2-1-5-14(12)15/h1-6,8,10-11,19H,7,9H2,(H,18,20)
InChI key
ZDAZUJBASMCUAK-UHFFFAOYSA-N
storage temp.
-80/-20
Purity
98%
Country of Origin
CN
Shipping Temperature
4
Physical Form
solid
Product Link
https://www.targetmol.com/compound/nicotredole
MSDS
https://www.targetmol.com/compound/nicotredole
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.