Skip to Content
Merck
All Photos(1)

Key Documents

PHL83937

Salicylic alcohol

new

phyproof® Reference Substance

Synonym(s):

2-Hydroxybenzyl alcohol, Diathesin, Salicain, Saligenol, Salicyl alcohol, Saligenin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH2OH
CAS Number:
Molecular Weight:
124.14
Beilstein:
1907195
MDL number:
UNSPSC Code:
12352200

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

manufacturer/tradename

PhytoLab

technique(s)

HPLC: suitable

mp

83-85 °C (lit.)

application(s)

food and beverages
pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

OCc1ccccc1O

InChI

1S/C7H8O2/c8-5-6-3-1-2-4-7(6)9/h1-4,8-9H,5H2

InChI key

CQRYARSYNCAZFO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service