Skip to Content
Merck
All Photos(1)

Key Documents

PHL22641

Deguelin

new

phyproof® Reference Substance

Synonym(s):

(−)-Deguelin, (7aS,13aS)-13,13a-Dihydro-9,10-dimethoxy-3,3-dimethyl-3H-bis[1]benzopyrano[3,4-b:6′,5′-e]pyran-7(7aH)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H22O6
CAS Number:
Molecular Weight:
394.42
MDL number:
UNSPSC Code:
12352200

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥98.0% (HPLC)

manufacturer/tradename

PhytoLab

technique(s)

HPLC: suitable

mp

85-87 °C (lit.)

application(s)

food and beverages
pharmaceutical

format

neat

storage temp.

−20°C

SMILES string

COc1cc2OC[C@H]3Oc4c(ccc5OC(C)(C)C=Cc45)C(=O)[C@H]3c2cc1OC

InChI

1S/C23H22O6/c1-23(2)8-7-12-15(29-23)6-5-13-21(24)20-14-9-17(25-3)18(26-4)10-16(14)27-11-19(20)28-22(12)13/h5-10,19-20H,11H2,1-4H3/t19-,20+/m1/s1

InChI key

ORDAZKGHSNRHTD-UXHICEINSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Inhibitor of activated Akt. Does not affect MAPK, ERK1/2, or JNK. Anticancer, chemoprotective agent.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service