Skip to Content
Merck
All Photos(1)

Key Documents

G005898

GC Analysis of Spearmint Essential Oil on SLB®-IL59

suitable for GC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

gas chromatography (GC): suitable

test parameters

column: SLB-IL59, 30 m x 0.25 mm I.D., 0.2 μm (28891-U)
oven: 50 °C, 3 °C/min to 300 °C
detector: FID
carrier gas: helium, 30 cm/sec, constant

application(s)

flavors and fragrances

Analysis Note

The volatile fraction of essential oils represents a quite complex matrix, being composed of a large variety of compounds, mainly terpenes, sesquiterpenes, alcohols and other oxygenated compounds, that cannot be all separeted in a single GC analysis. The ionic liquid GC column, SLB®-IL59 showed a good resolvling power toward all the classes of compounds present in the essential oils. It also provided good peak shape for both terpenes and oxygenated compounds thus proving it is a valuable polar GC column for the analyis of these types of samples. More than 50 compounds have been identified on this column by using mass spectra databases and by co-injection of standards.

Legal Information

SLB is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    α-Thujene
    α-Pinene
    Sabinene
    Myrcene
    α-Terpinene
    γ-Terpinene
    Terpinolene
    Eucalyptol
    p-Cymene
    Linalool
    Menthol
    α-Terpineol
    Menthyl acetate
    See All (13)

analytical column


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service