Skip to Content
Merck
All Photos(2)

Documents

SML3272

Sigma-Aldrich

Chroman 1

≥98% (HPLC)

Synonym(s):

(3S)-N-[2-[2-(Dimethylamino)ethoxy]-4-(1H-pyrazol-4-yl)phenyl]-3,4-dihydro-6-methoxy-2H-1-benzopyran-3-carboxamide, (3S)-N-[2-[2-(Dimethylamino)ethoxy]-4-(1H-pyrazol-4-yl)phenyl]-6-methoxy-3,4-dihydro-2H-chromene-3-carboxamide, (S)-Chroman 1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H28N4O4
CAS Number:
Molecular Weight:
436.50
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Highly potent and ROCK2-selective Rho kinase (ROCK1/2) inhibitor that is superior to Y-27632 in improving the viability of hPSCs in cultures.

Chroman 1 is a highly potent and ROCK2-selective Rho kinase inhibitor (ROCK1/2 IC50 = 52/1 pM) with much greater potency and selectivity than Y-27632 (ROCK1/2 IC50 = 71/46 nM). Chroman 1 is superior to Y-27632 in improving the viability of hPSCs in cultures (50 nM Chroman 1 is equivalent to 10 µM Y-27632; hESCs WA09 in E8 medium on vitronectin-coated plate). The combination of chroman 1, emricasan, polyamines (40 ng/ml putrescine, 4.5 ng/ml spermidine, and 8 ng/ml spermine), and trans-ISRIB (CEPT) is reported to enhance the survival of genetically stable hPSCs by simultaneously blocking several stress mechanisms that otherwise compromise cell structure and function.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Chen et al.
Nature methods, 18(5), 528-541 (2021-05-05)
Human pluripotent stem cells (hPSCs) are capable of extensive self-renewal yet remain highly sensitive to environmental perturbations in vitro, posing challenges to their therapeutic use. There is an urgent need to advance strategies that ensure safe and robust long-term growth

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service