Skip to Content
Merck
All Photos(1)

Documents

SML1461

Sigma-Aldrich

Kyotorphin acetate salt

≥98% (HPLC)

Synonym(s):

(2S)-2-[[(2S)-2-Amino-3-(4-hydroxyphenyl)propanoyl]amino]-5-(diaminomethylideneamino)pentanoic acid acetate salt, KTP, L-tyrosyl-L-arginine acetate salt, Tyr-Arg acetate salt, Tyrosyl-Arginine acetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23N5O4 · xC2H4O2
CAS Number:
Molecular Weight:
337.37 (free base basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

NC(NCCC[C@@H](C(O)=O)NC([C@@H](N)CC1=CC=C(O)C=C1)=O)=N.C

InChI

1S/C15H23N5O4.CH4/c16-11(8-9-3-5-10(21)6-4-9)13(22)20-12(14(23)24)2-1-7-19-15(17)18;/h3-6,11-12,21H,1-2,7-8,16H2,(H,20,22)(H,23,24)(H4,17,18,19);1H4/t11-,12-;/m0./s1

InChI key

KTMKVRFTDZZFFE-FXMYHANSSA-N

Biochem/physiol Actions

Kyotorphin is a neuroactive dipeptide (Tyr-Arg) shown to have analgesic, neuroprotective, and neuromodulatory properties. It is synthesized in nerve terminals and found in the brain and in cerebro-spinal fluid (CSF). Kyotorphin indirectly stimulates opioid receptors by releasing methionine enkephalin (met-enkephalin). CSF levels are lower in patients with persistent pain and in patients with Alzheimer′s disease.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service