Skip to Content
Merck
All Photos(1)

Documents

L2896

Sigma-Aldrich

Leucokinin I

≥97% (HPLC)

Synonym(s):

Asp-Pro-Ala-Phe-Asn-Ser-Trp-Gly-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H53N11O12
CAS Number:
Molecular Weight:
891.93
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

storage temp.

−20°C

Biochem/physiol Actions

Stimulates contraction of lower digestive tract

Other Notes

Neuropeptide originally isolated from head of Madeira cockroach Leucophaea maderae

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Holman, G.M., et al.
Comparative Biochemistry and Physiology. C, 84, 205-205 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service