Skip to Content
Merck
All Photos(1)

Documents

IST108078LR

ThermASeal Foil heat sealable seal

LabRoll, W × L 78 mm × 500 mm, pack of 1 roll

Synonym(s):

ThermASeal Foil heat sealable seal, bonding foil, non pierable bonding foil

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122410
NACRES:
NB.15

sterility

non-sterile

packaging

pack of 1 roll

manufacturer/tradename

IST Scientific IST-108-078LR

technique(s)

PCR: suitable

W × L

78 mm × 500 mm

suitability

suitable for PCR

Looking for similar products? Visit Product Comparison Guide

General description

Strong bonding foil to polypropylene, ideal for water thermal cyclers.

Features: Foil laminate, strong bonding, good solvent resistance including DMSO, peelable, non-pierceable.
Compatibility: Polypropylene (PP), Polystyrene (PS).
Seal integrity range: -200°C to 110°C.

Application

PCR, Ideal for water bath thermo cyclers, long term compound storage for hazardous materials

Legal Information

LabRoll is a trademark of IST Scientific
ThermASeal Foil is a trademark of IST Scientific

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service