Skip to Content
Merck
All Photos(1)

Documents

MABT849

Sigma-Aldrich

Anti-Myosin 7 (MYH7) Antibody, clone 4B51E8

clone 4B51E8, from rat

Synonym(s):

Myosin heavy chain 7, Myosin heavy chain slow isoform, MyHC-slow, Myosin heavy chain, Cardiac muscle beta isoform, MyHC-beta)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4B51E8, monoclonal

species reactivity

mouse

species reactivity (predicted by homology)

rat (based on 100% sequence homology), human (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... MYH7(4625)

Related Categories

General description

Myosin-7 (UniProt: Q91Z83; also known as Myosin heavy chain 7, Myosin heavy chain slow isoform, MyHC-slow, Myosin heavy chain, cardiac muscle beta isoform, MyHC-beta) is encoded by the MYH7 gene (Gene ID: 140781) in murine species. Myosins are actin-based motor molecules with ATPase activity essential for muscle contraction. Myosins form regular bipolar thick filaments that, together with actin thin filaments, constitute the fundamental contractile unit of skeletal and cardiac muscle. Myosin-7 is a component of thick filaments of the myofibrils. It contains actin- and ATP-binding sites in its conserved catalytic head domain. Its actin binding region is localized to amino acids 655-677 and 757-771 and the ATP-binding region is localized to amino acids 178-185. Myosin-7 has an N-terminal SH3-like domain (aa 32-81), a myosin motor domain (aa 85-778), and an IQ domain (aa 781-810). Muscle myosin is a hexameric protein that consists of 2 heavy chain subunits (MHC), 2 alkali light chain subunits (MLC) and 2 regulatory light chain subunits (MLC-2). Limited proteolysis of myosin heavy chain produces one light meromyosin (LMM) and one heavy meromyosin (HMM). HMM can be further cleaved into two globular sub fragments (S1) and 1 rod-shaped sub fragment (S2).

Specificity

Clone 4B51E8 is a rat monoclonal antibody that detects murine Myosin-7. It targets an epitope within 39 amino acids from the C-terminall half.

Immunogen

KLH-conjugated linear peptide corresponding to 39 amino acids from the C-terminal region of murine Mysoin-7.

Application

Anti-Myosin 7 (MYH7), clone 4B51E8, Cat. No. MABT849, is a rat monoclonal antibody that detects Mysoin-7 and has been tested for use in Immunofluorescence, Immunohistochemistry (Paraffin), and Western Blotting.
Immunohistochemistry (Paraffin) Analysis: A 1:25 dilution from a representative lot detected Myosin 7 (MYH7) in mouse skeletal muscle tissue.

Immunohistochemistry Analysis: A representative lot detected Myosin 7 (MYH7) in Immunohistochemistry applications (Sawano, S., et. al. (2016). PLoS One. 11(11):e0166080).

Immunofluorescence Analysis: A representative lot detected Myosin 7 (MYH7) in Immunofluorescence applications (Sawano, S., et. al. (2016). PLoS One. 11(11):e0166080).

Western Blotting Analysis: A representative lot detected Myosin 7 (MYH7) in Western Blotting applications (Sawano, S., et. al. (2016). PLoS One. 11(11):e0166080).

Western Blotting Analysis: 2 µg/mL from a representative lot detected Myosin 7 (MYH7) in mouse soleus muscle tissue lysate.

Quality

Evaluated by Immunohistochemistry (Paraffin) in mouse heart tissue.

Immunohistochemistry (Paraffin) Analysis: A 1:25 dilution of this antibody detected Myosin 7 (MYH7) in mouse heart tissue.

Target description

222.88 kDa calculated.

Physical form

Format: Purified

Storage and Stability

Stable for 1 year at 2-8°C from the date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service