Skip to Content
Merck
All Photos(1)

Documents

MABT371

Sigma-Aldrich

Anti-EB1, EB2 and EB3 Antibody, clone KT65

clone KT65, from rat

Synonym(s):

Microtubule-associated protein RP/EB family member 1, APC-binding protein EB1, End-binding protein 1, EB1, EB2, End binding protein 2, EB3, End binding protein 3, Microtubule-associated protein RP/EB family member 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

KT65, monoclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

Related Categories

General description

EB (End-binding) proteins are evolutionarily conserved and are important for extending tips of microtubules as well as regulating microtubule dynamics and communication with other intracellular substances. EB1 (End binding protein 1) encoded by the gene EB1/MAPRE1, EB2 (End binding protein 2) encoded by EB2/MAPRE2, and EB3 (End binding protein 3 also known as Microtubule-associated protein RP/EB family member 3) encoded by EB3/RP3/EBF3-S2/EBF3/MAPRE3 bind directly with end-tracking proteins (+TIPs). +TIPs influence microtubule growth, rescue, dynamicity, and suppression. EB1, EB2, and EB3 play major roles in cellular migration and adhesion, and are thought to associate with adenomatous polyposis coli tumor suppressor.

Specificity

This antibody is expected to detect EB1, EB2 and EB3.

Immunogen

GST-tagged recombinant protein corresponding to mouse EB1, EB2, and EB3.

Application

Immunocytochemistry Analysis: A representative lot detected EB1, EB2, and EB3 in NIH/3T3 cells (Van der Vaart, B., et al. (2011). J Cell Biol. 193(6):1083-1099).
Immunoprecipitation Analysis: A representative lot immunoprecipitated EB1, EB2, and EB3 in HeLa cell extracts (Van der Vaart, B., et al. (2011). J Cell Biol. 193(6):1083-1099).
Research Category
Cell Structure
Research Sub Category
Cytoskeletal Signaling
This Anti-EB1, EB2 & EB3 Antibody, clone KT65 is validated for use in western blotting, ICC & IP for the detection of EB1.

Quality

Evaluated by Western Blotting in MAPRE1 (EB1), MAPRE2 (EB2), and MAPRE3 (EB3) cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected EB1, EB2, and EB3 in 10 µg of MAPRE1 (EB1), MAPRE2 (EB2), and MAPRE3 (EB3) cell lysate.

Target description

31, 37, 32 kDa calculated. The calculated MW of EB1 (31 kDa), EB2 (37 kDa), and EB3 (32 kDa). The recombinants used to test the reactivity of this antibody to EB1, EB2, and EB3 have the following MW′s. EB1 (56 kDa), EB2 (62 kDa), and EB3 (58 kDa).

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ in buffer containing PBS.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service