Skip to Content
Merck
All Photos(1)

Key Documents

ABD122

Sigma-Aldrich

Anti-Dishevelled Antibody

serum, from rabbit

Synonym(s):

DSH, DVL1, DVL2, DVL3, DSH Homolog 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.75

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... DVL3(1857)

General description

Dishevelled (DVL1), the human homolog of the Drosophila dishevelled gene (dsh) encodes a cytoplasmic phosphoprotein that regulates cell proliferation. The protein acts as a transducer molecule for developmental processes, including segmentation and neuroblast specification. Dishevelled plays a role both in canonical and non-canonical Wnt signaling. DVL1 mutation is likely responsible for transformation in neuroblastomas, and the Schwartz-Jampel syndrome and Charcot-Marie-Tooth disease type 2A have been mapped to the same region as DVL1 gene, thus Dishevelled maybe important in these diseases as well.

Specificity

Recognizes Human, Mouse and Rat Dishevelled 1, 2 and 3.

Immunogen

GST-tagged recombinant protein corresponding to Human Dishevelled.

Application

Immunocytochemistry Analysis: A representative lot was used by an independent laboratory in PC12 cells. (Kishida et al., 2007. Dvl regulates endo- and exocytotic processes through binding to synaptotagmin. Genes to Cells, 12, 49–61).
Research Category
Stem Cell Research
Research Sub Category
Developmental Signaling
This Anti-Dishevelled Antibody is validated for use in Western Blotting and Immunocytochemistry for the detection of Dishevelled.

Quality

Evaluated by Western Blot in HEK293T cell lysate.

Western Blot Analysis: A 1:1000 of this antibody detected Dishevelled in 10 µg of HEK293T cell lysate.

Target description

~ 81 kDa observed

Physical form

Rabbit polyclonal serum containing 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anja Bufe et al.
Proceedings of the National Academy of Sciences of the United States of America, 118(34) (2021-08-22)
Canonical Wnt signaling plays critical roles in development and tissue renewal by regulating β-catenin target genes. Recent evidence showed that β-catenin-independent Wnt signaling is also required for faithful execution of mitosis. However, the targets and specific functions of mitotic Wnt

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service