Skip to Content
Merck
All Photos(2)

Key Documents

8.43567

Sigma-Aldrich

N,N,N′,N′-Tetramethyl-1,3-propanediamine

for synthesis

Synonym(s):

N,N,N′,N′-Tetramethyl-1,3-propanediamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H18N2
CAS Number:
Molecular Weight:
130.23
MDL number:
UNSPSC Code:
12352116
EC Index Number:
203-818-8

vapor pressure

6.5 hPa ( 20 °C)

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

180 °C (DIN 51794)

potency

620 mg/kg LD50, oral (Rat)
1180 mg/kg LD50, skin (Rat)

expl. lim.

2.9-22.5 % (v/v)

pH

12.5 (20 °C, 100 g/L in H2O)

mp

<-80 °C

transition temp

flash point 31 °C (DIN 51755 Part 1)

density

0.78 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C7H18N2/c1-8(2)6-5-7-9(3)4/h5-7H2,1-4H3/p+2

InChI key

DMQSHEKGGUOYJS-UHFFFAOYSA-P

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.783 - 0.787
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

87.8 °F - Equilibrium method

Flash Point(C)

31 °C - Equilibrium method


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service