Skip to Content
Merck
All Photos(1)

Key Documents

1.28239

SAFC

Zinc sulfate heptahydrate

new

EMPROVE® API, Ph. Eur., BP, ChP, JP, USP

Pharma Manufacturing

Synonym(s):

Zinc sulfate heptahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
O4SZn · 7H2O
CAS Number:
Molecular Weight:
287.56
UNSPSC Code:
12352200

Agency

BP
ChP
JP
Ph. Eur.
USP

Quality Level

product line

EMPROVE® API

application(s)

clinical nutrition
liquid formulation
pharma/biopharma processes
pharma/biopharma processes
pharmaceutical

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

We provide industry-leading production know-how and comprehensive regulatory support for high-quality APIs, backed by nearly 200 years of experience. From a complete portfolio of innovative folates to custom synthesis, our customized and scalable solutions meet your specific production needs. As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Zinc sulfate is a mineral salt broadly used in pharmaceutical formulations and cosmetics.

Packaging

For global market (except China) 1282390003 (3X1kg); 1282391001 (1 kg); 1282398026 (25 kg); 1282398029 (25kg)
For China market 1282390103 (3X1kg); 1282391101 (1kg);1282398126 (25kg); 1282398129 (25kg)
For global market (except China) 1282390003 (plastic bottle in sample kit box); 1282391001 (plastic bottle); 1282398026 (Double PE bag in plastic drum); 1282398029 (Double PE bag in carton box)
For China market 1282390103 (plastic bottle in sample kit box); 1282391101 (plastic bottle);1282398126 (Double PE bag in plastic drum); 1282398129 (Double PE bag in carton box)

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service