Skip to Content
Merck
All Photos(1)

Key Documents

1.07807

Supelco

Tin

fine powder (particle size < 71 µm) EMPLURA®

Synonym(s):

Tin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sn
CAS Number:
Molecular Weight:
118.71
MDL number:
UNSPSC Code:
12141745
EC Index Number:
231-141-8
NACRES:
NA.21
form:
solid

Quality Level

product line

EMPLURA®

form

solid

bp

2260 °C/1013 hPa

mp

232 °C

density

7.3 g/cm3 at 20 °C

cation traces

Bi: ≤0.02%
Cu: ≤0.01%
Fe: ≤0.02%
Pb: ≤0.05%
Sb: ≤0.02%

storage temp.

2-30°C

InChI

1S/Sn

InChI key

ATJFFYVFTNAWJD-UHFFFAOYSA-N

Analysis Note

Bi (Bismuth): ≤ 0.02 %
Cu (Copper): ≤ 0.01 %
Fe (Iron): ≤ 0.02 %
Pb (Lead): ≤ 0.05 %
Sb (Antimony): ≤ 0.02 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service