Skip to Content
Merck
All Photos(1)

Key Documents

KEY458251057

Key Organics / BIONET

1-benzyl-6-oxo-1,6-dihydropyridine-3-carbohydrazide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
243.27
MDL number:

Iupac Name

1-benzyl-6-oxo-1,6-dihydro-3-pyridinecarbohydrazide

Inchi Code

1S/C13H13N3O2/c14-15-13(18)11-6-7-12(17)16(9-11)8-10-4-2-1-3-5-10/h1-7,9H,8,14H2,(H,15,18)

InChI key

BEIWEJMLXMJIED-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/10E-384S

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

MSDS

https://www.keyorganics.net/sds/10E-384S


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library