JWPH3249C651
J & W Pharmlab LLC
4-(Tetrahydro-pyran-4-yl)-benzoic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
4-tetrahydro-2H-pyran-4-ylbenzoic acid
Inchi Code
1S/C12H14O3/c13-12(14)11-3-1-9(2-4-11)10-5-7-15-8-6-10/h1-4,10H,5-8H2,(H,13,14)
InChI key
UOTCSFXNJIBLLZ-UHFFFAOYSA-N
Purity
96%
Country of Origin
US
Physical Form
Off White Solid
Product Link
https://www.jwpharmlab.com/home/product/15R0100
MSDS
https://www.jwpharmlab.com/home/product/15R0100
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.