Skip to Content
Merck
All Photos(1)

Key Documents

ENAH97A1849E

Enamine

1-bromo-3-isocyanatopropane

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
164
MDL number:

Iupac Name

1-bromo-3-isocyanatopropane

Inchi Code

1S/C4H6BrNO/c5-2-1-3-6-4-7/h1-3H2

InChI key

KMDXMJQBNOVBNE-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-4271421

MSDS

https://www.enaminestore.com/catalog/EN300-4271421


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library