Skip to Content
Merck
All Photos(1)

Key Documents

ENAH0424F67F

Enamine

tert-butyl 3-hydroxy-3-(2-oxopropyl)azetidine-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
229.28
MDL number:

Iupac Name

tert-butyl 3-hydroxy-3-(2-oxopropyl)azetidine-1-carboxylate

Inchi Code

1S/C11H19NO4/c1-8(13)5-11(15)6-12(7-11)9(14)16-10(2,3)4/h15H,5-7H2,1-4H3

InChI key

MBRMXIXLBIZMPR-UHFFFAOYSA-N

storage temp.

4

Purity

92

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

oil

Product Link

https://www.enaminestore.com/catalog/EN300-1622914

Pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.enaminestore.com/catalog/EN300-1622914


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library