ENA514234648
Enamine
(4-cyclopropyl-1-methyl-1H-pyrazol-5-yl)methanamine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
(4-cyclopropyl-1-methyl-1H-pyrazol-5-yl)methanamine
Inchi Code
1S/C8H13N3/c1-11-8(4-9)7(5-10-11)6-2-3-6/h5-6H,2-4,9H2,1H3
InChI key
FFKCXKUTCZAYDH-UHFFFAOYSA-N
storage temp.
4
Purity
95
Country of Origin
UA
Shipping Temperature
Normal
Product Link
https://www.enaminestore.com/catalog/EN300-269587
Signal Word
Danger
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-269587
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.