mp
162-163
Iupac Name
[1,1':4',1''-terphenyl]-2-amine
Inchi Code
1S/C18H15N/c19-18-9-5-4-8-17(18)16-12-10-15(11-13-16)14-6-2-1-3-7-14/h1-13H,19H2
InChI key
LZCYDNMVXHWZSD-UHFFFAOYSA-N
storage temp.
RT
Purity
95
Country of Origin
UA
Shipping Temperature
Normal
Physical Form
powder
Product Link
https://www.enaminestore.com/catalog/EN300-71996
Melting Point
162-163
MSDS
https://www.enaminestore.com/catalog/EN300-71996
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.