Skip to Content
Merck
All Photos(1)

Key Documents

ENA238710433

Enamine

1-(3-methoxyphenyl)imidazolidine-2,4,5-trione

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
220.18
MDL number:

Iupac Name

1-(3-methoxyphenyl)-2,4,5-imidazolidinetrione

Inchi Code

1S/C10H8N2O4/c1-16-7-4-2-3-6(5-7)12-9(14)8(13)11-10(12)15/h2-5H,1H3,(H,11,13,15)

InChI key

NIQKUAZKRJCIOQ-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-34803

MSDS

https://www.enaminestore.com/catalog/EN300-34803


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library