COMH95E96277
Combi-Blocks, Inc.
4-Bromo-1-tert-butyl-6-fluoroindazole
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
4-bromo-1-(tert-butyl)-6-fluoro-1H-indazole
Inchi Code
1S/C11H12BrFN2/c1-11(2,3)15-10-5-7(13)4-9(12)8(10)6-14-15/h4-6H,1-3H3
InChI key
UQOUVTXRCBJRMV-UHFFFAOYSA-N
storage temp.
Refrigerated
Purity
96%
Country of Origin
CN
Product Link
https://www.combi-blocks.com/cgi-bin/find.cgi?IN-0616
MSDS
https://www.combi-blocks.com/cgi-bin/find.cgi?IN-0616
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.