COM448619298
Combi-Blocks, Inc.
Methyl 4-chloro-7,8-dimethylquinoline-2-carboxylate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
methyl 4-chloro-7,8-dimethyl-2-quinolinecarboxylate
Inchi Code
1S/C13H12ClNO2/c1-7-4-5-9-10(14)6-11(13(16)17-3)15-12(9)8(7)2/h4-6H,1-3H3
InChI key
VZYCKZKXAVZPOB-UHFFFAOYSA-N
Purity
98%
Country of Origin
CN
Product Link
https://www.combi-blocks.com/cgi-bin/find.cgi?HC-2323
MSDS
https://www.combi-blocks.com/cgi-bin/find.cgi?HC-2323
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.